Anti-tubulin beta-5 Rabbit Monoclonal Antibody [clone: 9F3] (Alexa Fluor 555)
Catalog # CSTA2116S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:tubulin beta-5
- Clonality:Monoclonal
- Clone:9F3
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Zebrafish,Monkey,Human,Bovine,Rat,Mouse
- Sequence:CSDEHGIDPTGTYHGDSDLQLDRI
- Storage temperature:4 °C
- Shipping temperature:Cold
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: tubulin beta-5
Clonality: Monoclonal
Clone: 9F3
Conjugation: Alexa Fluor 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey;Zebrafish;Bovine