Anti-SHIP2 (Tyr986/Tyr987) phosphate Rabbit Polyclonal Antibody
CSTA2008S
- Antibody type:Primary
- Antigen symbol:SHIP2 (Tyr986/Tyr987) phosphate
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Sequence:CNSFNNPAyyVLEGVP
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: SHIP2 (Tyr986/Tyr987) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human