Ethyl propiolate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:623-47-2
:
Formula:C₅H₆O₂
MDL Number:MFCD00009184
Boiling Pt:120 °C (1013 hPa)
Melting Pt:9 °C
Density:0.9788 g/cm³ (4 °C)
ADR:3,III
:M-Clarity™ quality level = MQ100
:98.1 g/mol
EINECS:210-795-8
UN:3272
Frequently Bought Together