Methyl phenylpropiolate, Sigma-Aldrich®
Catalog # SIAL543195-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:160.17 g/mol
Boiling Pt:248 °C (1013 hPa)
Melting Pt:26 °C
Density:1.083 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
CAS Number:4891-38-7
Formula:C₁₀H₈O₂
MDL Number:MFCD00041685