Cesium formate, Sigma-Aldrich®
Catalog # SIAL433764-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:177.92 g/mol
:M-Clarity™ quality level = MQ100
EINECS:222-492-8
CAS Number:3495-36-1
Formula:CHCsO₂
MDL Number:MFCD00039103