1-Butyl-3-methylimidazoliumtetrafluoroborate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:174501-65-6
:
Formula:C₈H₁₅BF₄N₂
MDL Number:MFCD03095449
:226.02 g/mol
Boiling Pt:∼200 °C (1013 hPa)
Melting Pt:–71 °C
Density:1.2 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:>110 °C
Frequently Bought Together