Chromium hexacarbonyl, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13007-92-6
:
Formula:C₆CrO₆
MDL Number:MFCD00010945
:220.06 g/mol
Merck Index:13,02253
Boiling Pt:210 °C (1013 hPa)
Melting Pt:90 °C
Density:1.77 g/cm³ (20 °C)
EINECS:235-852-4
UN:3466
ADR:6.1,II
Frequently Bought Together