2-Aminobenzothiazole, Sigma-Aldrich®
: Merck
: Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II
- :136-95-8
- :C₇H₆N₂S
- :MFCD00005785
- :150.2 g/mol
- :13,00423
- :190…195 °C (0.05 mmHg)
- :127…130 °C
- :M-Clarity™ quality level = MQ100
- :205-268-4
: Benzothiazol-2-ylamine, Benzo[d]thiazol-2-amine, Cerium(III)-Ionophore II
- :136-95-8
- :C₇H₆N₂S
- :MFCD00005785
- :150.2 g/mol
- :13,00423
- :190…195 °C (0.05 mmHg)
- :127…130 °C
- :M-Clarity™ quality level = MQ100
- :205-268-4
Product Family Options
- Pack typePk