



Aniline hydrochloride
Catalog # ROTH5113.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
Specifications
About this item
:
Merck Index:13,00616
Boiling Pt:245 °C (1013 hPa)
Melting Pt:198 °C
Density:1.2215 g/cm³ (20 °C)
Flash Pt:193 °C
EINECS:205-519-8
UN:1548
ADR:6.1,III
:GHS06
:GHS05
:GHS08
:GHS09
:129.59 g/mol
CAS Number:142-04-1
Formula:C₆H₈ClN
MDL Number:MFCD00012958