Water
Catalog # OXOIBO0209M
Supplier: THERMO FISHER DIAGNOSTICS
CAS Number:
Specifications
- Pk:10
- Seq:0010
Specifications
About this item
:
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332