Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

2-Bromothiazole

Glass bottle for solids, 25 g
Catalog # MOLE18922792-25G
Marketsource
Supplier:   Molekula
2-Bromothiazole
Catalog # MOLE18922792-25G
Supplier:  Molekula
Marketsource
CAS Number:  3034-53-5
2-Bromothiazole

Glass bottle for solids, 25 g

CAS Number:  3034-53-5
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:3034-53-5
  • Formula:C₃H₂BrNS
  • MDL Number:MFCD00005316
  • Molecular Weight:164.03 g/mol
  • Boiling Pt:171 °C (1013 hPa)
  • Density:1,82 g/cm³ (20 °C)
  • EINECS:221-229-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Cat. No.:
    MOLE18922792-25G

Specifications