Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
2-Thiouracil

2-Thiouracil

Glass bottle for solids, 100 g
Catalog # MOLE44036154-100G
Marketsource
Supplier:   Molekula
2-Thiouracil
Catalog # MOLE44036154-100G
Supplier:  Molekula
Marketsource
CAS Number:  141-90-2
2-Thiouracil

Glass bottle for solids, 100 g

CAS Number:  141-90-2
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  4-Hydroxypyrimidine-2(1H)-thione
  • CAS Number:141-90-2
  • Formula:C₄H₄N₂OS
  • MDL Number:MFCD00006039
  • Molecular Weight:128.15 g/mol
  • Merck Index:12,09504
  • Melting Pt:340 °C
  • EINECS:205-508-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications