2-Amino-6-nitrobenzothiazole
Catalog # MOLE49445000-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Melting Pt:252 °C
Density:1.649 g/cm³ (20 °C)
UN:2811
ADR:6.1,III
:195.2 g/mol
EINECS:228-513-7
CAS Number:6285-57-0
Formula:C₇H₅N₃O₂S
MDL Number:MFCD00005786