

Sodium bromate, Msynth® plus, Sigma-Aldrich®
Catalog # 8.45066.9999
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
Specification Test Results
- Solubility:364 g/l (20 °C)
- Melting Point:381 °C (decomposition)
- Molar Mass:150.89 g/mol
- Density:3.3 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral,Aqueous solution
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
About this item
:
:150.89 g/mol
Merck Index:12,08736
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:381 °C
Density:3.254 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-160-4
UN:1494
ADR:5.1,II
:M-Clarity™ quality level = MQ400
:GHS03
:GHS07
CAS Number:7789-38-0
Formula:BrNaO₃
MDL Number:MFCD00003476