
Trimethylammonium chloride for synthesis, Sigma-Aldrich®
: Merck
:593-81-7
:
:C₃H₁₀ClN
:MFCD00012478
:95.57 g/mol
:12,09841
:273…278 °C
:∼1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:209-810-0
:GHS07
:Ambient
- Solubility:(20 °C) soluble
- Melting Point:273 - 278 °C
- Molar Mass:95.57 g/mol
- pH:5 (100 g/l, H2O, 20 °C)
Frequently Bought Together