Order Entry
United Kingdom
ContactUsLinkComponent
Sodium chloroacetate for synthesis, Sigma-Aldrich®
Toxic
Environment
Sodium chloroacetate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Environment
Sodium chloroacetate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    820 g/l (20 °C)
  • Molar Mass:
    116.47 g/mol
  • Bulk Density:
    850 kg/m3
  • Flash Point:
    270 °C
  • pH:
    4.5 - 9.0 (50 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together