Butyl vinyl ether stabilised for synthesis, Sigma-Aldrich®
: Merck
: n-Butyl vinyl ether
:111-34-2
:
:C₆H₁₂O
:MFCD00009454
:94
:0.7804 g/cm³ (20 °C)
:100 ppm Stabilised with potassium hydroxide
:100.16 g/mol
:–91.9 °C
:2352
:3 II
:M-Clarity™ quality level = MQ200
:203-860-7
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:3 g/l (20 °C)
- Melting Point:-113 °C
- Molar Mass:100.16 g/mol
- Boiling Point:93 - 94 °C (1013 hPa)
- Vapor Pressure:51 hPa (20 °C)
- Flash Point:-6 °C DIN 51755 Part 1
- Refractive Index:1.401 (20 °C, 589 nm)
- Explosion Limit:1.2 - 8.6 %(V)
- Density:0.78 g/cm3 (20 °C)
- Ignition Point:190 °C DIN 51794
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Frequently Bought Together