
Nitric acid 1 M HNO3 (1N) volumetric solution, Fluka™
Catalog # 35315-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
:63.01 g/mol
Boiling Pt:100 °C
Density:1.03 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349