ω-Bromo-4-nitroacetophenone
Marketsource
Supplier: FLUOROCHEM
Jump to:
About this item
Synonyms: α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone
- CAS Number:99-81-0
- Formula:C₈H₆BrNO₃
- MDL Number:MFCD00007356
- Molecular Weight:244.04 g/mol
- Melting Pt:100 °C
- Storage Temperature:Ambient
- EINECS:202-789-9
2 Options Available Below
Product Details & Documents
Synonyms: α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone
- CAS Number:99-81-0
- Formula:C₈H₆BrNO₃
- MDL Number:MFCD00007356
- Molecular Weight:244.04 g/mol
- Melting Pt:100 °C
- Storage Temperature:Ambient
- EINECS:202-789-9
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



