3'-Chloropropiophenone
Catalog # FLUO003621-25G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:168.62 g/mol
Boiling Pt:249 °C (1013 hPa)
Melting Pt:43…47 °C
EINECS:252-242-3
CAS Number:34841-35-5
Formula:C₉H₉ClO
MDL Number:MFCD00009925