£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
2-Chlorobenzyl alcohol
2-Chlorobenzyl alcohol
2-Chlorobenzyl alcohol
FLUO001375-100G
:
FLUOROCHEM
:
2-Chlorobenzyl alcohol
FLUO001375-100G
:
FLUOROCHEM
:
Pk:
100 g
Seq:
3401
:
:
227 °C (1013 hPa)
:
68…71 °C
:
142.58 g/mol
:
241-801-7
:
17849-38-6
:
C₇H₇ClO
:
MFCD00004604