(2,3-Dichlorophenoxy)acetic acid
Catalog # EHERC11939000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
Boiling Pt:329 °C (1013 hPa)
Melting Pt:195…198 °C
:221.04 g/mol
EINECS:221-022-9
CAS Number:2976-74-1
Formula:(C₆H₃Cl₂)OCH₂COOH
MDL Number:MFCD00004299