Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2,3,3-Tetrafluoro-1,4-butanediol
2,2,3,3-Tetrafluoro-1,4-butanediol ≥97%
2,2,3,3-Tetrafluoro-1,4-butanediol ≥97%
2,2,3,3-Tetrafluoro-1,4-butanediol ≥97%
L16635.06
:
Thermo Fisher Scientific
:
2,2,3,3-Tetrafluoro-1,4-butanediol ≥97%
L16635.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
425-61-6
:
:
C₄H₆F₄O₂
:
MFCD00042375
:
162.08 g/mol
:
110…112 °C (13 mmHg)
:
80…87 °C
:
GHS07
:
Ambient