Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

2-(4-Fluorophenyl)ethanol ≥97%

2-(4-Fluorophenyl)ethanol ≥97%
Certificates
2-(4-Fluorophenyl)ethanol ≥97%
2-(4-Fluorophenyl)ethanol ≥97%

2 Options Available Below

Product Details & Documents

Purity: 97% CAS No.: 7589-27-7 Molecular Formula: C8H9FO Formula Weight: 140.16 Flash Point: 104°(219°F) Refractive Index: 1.5065 Boiling Point: 118°/20mm Beilstein Reference: 2354310 MDL No.: MFCD00002898

Synonyms:  4-Fluorophenethyl alcohol
  • CAS Number:7589-27-7
  • Formula:C₈H₉FO
  • MDL Number:MFCD00002898
  • Molecular Weight:140.16 g/mol
  • Boiling Pt:118 °C (20 mmHg)
  • Density:1.121
  • Flash Pt:104 °C (219 °F)
  • Storage Temperature:Ambient
  • EINECS:231-499-5
Product Family Options

Product Information