Indole-5-carbaldehyd ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 1196-69-6 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 96-100° MDL No.: MFCD02093664
Synonyms: Indole-5-carboxaldehyde, 1H-Indole-5-carboxaldehyde
- CAS Number:1196-69-6
- Formula:C₉H₇NO
- MDL Number:MFCD02093664
- Molecular Weight:145.16 g/mol
- Melting Pt:96…100 °C
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 1196-69-6 Molecular Formula: C9H7NO Formula Weight: 145.16 Melting Point: 96-100° MDL No.: MFCD02093664
Synonyms: Indole-5-carboxaldehyde, 1H-Indole-5-carboxaldehyde
- CAS Number:1196-69-6
- Formula:C₉H₇NO
- MDL Number:MFCD02093664
- Molecular Weight:145.16 g/mol
- Melting Pt:96…100 °C
- Storage Temperature:Ambient
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



