Methyl phenylpropiolate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4891-38-7
:
Formula:C₁₀H₈O₂
MDL Number:MFCD00041685
:160.17 g/mol
Boiling Pt:109…110 °C (2 mmHg)
Melting Pt:26 °C
Density:1.083
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
Frequently Bought Together