Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
3,3'-Thiodipropionic acid ≥98%

3,3'-Thiodipropionic acid ≥98%

Irritant
3,3'-Thiodipropionic acid ≥98%
Certificates
3,3'-Thiodipropionic acid ≥98%
3,3'-Thiodipropionic acid ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 111-17-1 Molecular Formula: C6H10O4S Formula Weight: 178.21 Melting Point: 127-134° Beilstein Reference: 1210299 Merck Reference: 14,9332 MDL No.: MFCD00002781

Synonyms:  3,3-Thiodipropionic acid
  • CAS Number:111-17-1
  • Formula:C₆H₁₀O₄S
  • MDL Number:MFCD00002781
  • Molecular Weight:178.21 g/mol
  • Merck Index:14,9332
  • Boiling Pt:347 °C (1013 hPa)
  • Melting Pt:127…134 °C
  • Density:1.496 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-841-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - £25.50
    £25.50
    1 * 250 g
       £25.50

  • Glass bottle for solids
    1.000 g
    Log in for availability
    Log in for availability
    1 * 1.000 g
    1 * 1.000 g - £48.70
    £48.70
    1 * 1.000 g
       £48.70

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.