4,4'-Dihydroxybenzophenone ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 611-99-4 Molecular Formula: C13H10O3 Formula Weight: 214.22 Melting Point: 221-225° Beilstein Reference: 1874572
Synonyms: Bis(4-hydroxyphenyl) ketone
- CAS Number:611-99-4
- Formula:C₁₃H₁₀O₃
- MDL Number:MFCD00002358
- Molecular Weight:214.22 g/mol
- Boiling Pt:375 °C (1013 hPa)
- Melting Pt:221…225 °C
- Density:1.336 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:210-288-1
3 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 611-99-4 Molecular Formula: C13H10O3 Formula Weight: 214.22 Melting Point: 221-225° Beilstein Reference: 1874572
Synonyms: Bis(4-hydroxyphenyl) ketone
- CAS Number:611-99-4
- Formula:C₁₃H₁₀O₃
- MDL Number:MFCD00002358
- Molecular Weight:214.22 g/mol
- Boiling Pt:375 °C (1013 hPa)
- Melting Pt:221…225 °C
- Density:1.336 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:210-288-1



