

Thulium(III) nitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36548-87-5
:
Formula:N₃O₉Tm·xH₂O
MDL Number:MFCD00149875
ADR:5.1,III
:354.95 g/mol (anhy)
EINECS:239-069-9
UN:1477
:GHS03
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together