1,3-Dioxolane 99.5+% stabilised, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:646-06-0
:
Formula:C₃H₆O₂
MDL Number:MFCD00003207
:74.08 g/mol
Boiling Pt:75 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.06 g/cm³
Flash Pt:–6 °C
Storage Temperature:Ambient
EINECS:211-463-5
UN:1166
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3990 to 1.4010 (20°C, 589 nm)
- GC:≥99.50 %
- Stabilizer:60 to 100 ppm BHT
Specification Test Results
Frequently Bought Together