
Tetrapotassium diphosphate 98%, extra pure
:7320-34-5
:
:K₄O₇P₂
:MFCD00011393
:330.34 g/mol
:12,07833
:? 1500 °C (1013 hPa)
:1090 °C
:1,1 g/cm³ (20 °C)
:230-785-7
:GHS07
:Ambient
- Appearance:White Crystalline powder
- Titration with HClO4:97.5 to 102.5 %
- Chloride (Cl):≤100 ppm
- Sulfate (SO4):≤20 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
Frequently Bought Together