Order Entry
United Kingdom
ContactUsLinkComponent
2-(4-Bromophenyl)ethanol 99%
Irritant
2-(4-Bromophenyl)ethanol 99%
Catalog # ACRO179590100
CAS Number:  
undefined
Irritant
2-(4-Bromophenyl)ethanol 99%
Catalog # ACRO179590100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5720 to 1.5740 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

About this item

 :
 :201.06 g/mol
Boiling Pt:96…99 °C (.5 mmHg)
Melting Pt:36…38 °C
Density:1.43 g/cm³
Flash Pt:>110 °C
EINECS:225-093-7
 :GHS07
CAS Number:4654-39-1
Formula:C₈H₉BrO
MDL Number:MFCD00002897