


(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-47-9
:
Formula:C₅H₁₀O
MDL Number:MFCD00005367
:86.13 g/mol
Boiling Pt:78…80 °C (760 mmHg)
Melting Pt:–136 °C
Density:0.86 g/cm³
Flash Pt:–11 °C
Storage Temperature:Ambient
EINECS:202-507-4
UN:2536
ADR:3,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4046 to 1.4066 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥99.0 %
- Stabilizer:150 to 400 ppm BHT
Specification Test Results
Frequently Bought Together