Order Entry
United Kingdom
ContactUsLinkComponent
α-Bromo-m-toluonitrile 95%
α-Bromo-m-toluonitrile 95%
Certificates
undefined
α-Bromo-m-toluonitrile 95%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    92 to 97 °C
  • GC:
    ≥94 %

Specification Test Results

Related Information

Frequently Bought Together