Ethyl levulinate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:539-88-8
:
Formula:C₇H₁₂O₃
MDL Number:MFCD00009209
:144.17 g/mol
Merck Index:13,03852
Boiling Pt:93 °C (1013 hPa)
Density:1.012 g/cm³
Flash Pt:90 °C
EINECS:208-728-2
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4210 to 1.4240 (20°C, 589 nm)
- Density:(20°C) 1.009 to 1.014 g/ml
- GC:≥97.5 %
- Acid value:≤5 mg KOH/g
Specification Test Results
Frequently Bought Together