1-Amino-2-methylnaphthalene
Catalog # ABCRAB136336.0005
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
:
:157.22 g/mol
CAS Number:2246-44-8
Formula:C₁₁H₁₁N
MDL Number:MFCD00059135