
4-Amino-5-(2-furyl)-4H-1,2,4-triazole-3-thiol
Catalog # APOSOR928166-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:182.2 g/mol
Melting Pt:205 - 206 °C
:GHS07
CAS Number:80809-38-7
Formula:C₆H₆N₄OS
MDL Number:MFCD03044388
Melting Pt:205…206 °C