
2-Bromo-4-chloro-6-fluorobenzoyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1805575-89-6
:
Formula:C₇H₂BrCl₂FO
MDL Number:MFCD28733936
:271.9 g/mol
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together
4-Methyl-2-pentanone 99+% for HPLC
Supplier: Thermo Fisher Scientific
11 components: Ca 2mg/l Zr 5mg/l K 5mg/l Zn 5mg/l Na 5mg/l Fe 5mg/l Al 5mg/l Ti 5mg/l Si 5mg/l P 10mg/l Mn 5mg/l in HNO3 5% 1 * 1 L
Supplier: CUSTOM MADE CHEMICALS LAB
3-(Chloromethyl)benzoyl chloride 98%
Supplier: Thermo Fisher Scientific
Butyl acetate ≥99%
Supplier: Thermo Fisher Scientific
Methyl methanesulphonate 99%
Supplier: Thermo Fisher Scientific
Ethanol 90% 90% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
N,N-Dimethylacetamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Methylbutane ≥99%, Reagent Grade
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Diisopropyl ether stabilised for analysis
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95% for residue analysis, for trace analysis of polyaromatic hydrocarbons
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone ACS
Supplier: Thermo Fisher Scientific
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific