Isonicotinic acid-N-oxide 98%
:13602-12-5
:
:C₆H₅NO₃
:MFCD00006209
:139.11 g/mol
:469/760 mm °C
:270 °C
:1.34
:237.4 °C
:GHS07
:Refrigerator
Frequently Bought Together
Methyl ethyl ketone ≥99%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
: VWR Chemicals
Glycerine ≥86-88% (w/w) in aqueous solution for analysis
: Thermo Fisher Scientific
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
: Thermo Fisher Scientific