Ethylene carbonate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:96-49-1
:
Formula:C₃H₄O₃
MDL Number:MFCD00005382
:88.06 g/mol
Boiling Pt:243…244/740 mm °C
Melting Pt:35…38 °C
Density:1.321
Flash Pt:143 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together