Pesticide reference standard, 2,4,6-Trichloroanisole 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL17714600IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:87-40-1
- Description:2,4,6-Trichloroanisole 10 µg/ml in isooctane
- Formula:C₇H₅Cl₃O
- Description:2,4,6-Trichloroanisole