(±)-4-Methylcyclohexene, Sigma-Aldrich®
Catalog # SIALM39008-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:96.17 g/mol
Boiling Pt:103 °C (1013 hPa)
Melting Pt:–115.5 °C
Density:0.7991 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-715-4
UN:3295
ADR:3,II
:M-Clarity™ quality level = MQ100
CAS Number:591-47-9
Formula:C₇H₁₂
MDL Number:MFCD00001573