2-Chloro-α-methylbenzyl alcohol 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13524-04-4
:
Formula:C₈H₉ClO
MDL Number:MFCD00041037
:156.61 g/mol
Boiling Pt:121…123 °C (14 mmHg)
Storage Temperature:Ambient
EINECS:236-868-4
Frequently Bought Together