APC-iFluor™ 750 Tandem, ReadiUse™ fluorescent probe, preactivated
Catalog # AATB2571
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 mg
- Seq:2070
- Environmentally Preferable:
Specifications
About this item
:
:105000 g/mol
ADR:5,II
:GHS07