£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United Kingdom
ContactUsLinkComponent
Quin 2-Am
Quin 2-Am ≥95%
Quin 2-Am ≥95%
AATB21050
:
AAT BIOQUEST
:
Quin 2-Am ≥95%
AATB21050
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
829.765 g/mol
:
5,II
:
GHS07
:
83104-85-2
:
C₃₈H₄₃N₃O₁₈