Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
EGTA, AM
EGTA, AM ≥90%
EGTA, AM ≥90%
EGTA, AM ≥90%
Catalog # AATB21005
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
EGTA, AM ≥90%
Catalog # AATB21005
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
:
668.598 g/mol
ADR:
5,II
:
GHS07
CAS Number:
99590-86-0
Formula:
C₂₆H₄₀N₂O₁₈
Product Details