Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Semicarbazide (SEM)
AATB50008
:
- Pack type:Vial
- Pk:1 mg
- Conjugation:Semicarbazide (SEM)
- Protein/peptide type:Native
- Source:Serum
- Species:Megathura crenulata
- Environmentally Preferable:
- Protein synonyms:KLH
- Protein/peptide name:Keyhole limpet hemocyanin
Semicarbazide (SEM)-KLH Conjugate is used for developing anti-Semicarbazide (SEM) antibodies that are typically used in an ELISA assay.
- High immunogenicity
- Useful for antibody development
- ELISA assay building block
: For research use only.