Deuterium oxide (99.96% D) for NMR spectroscopy
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:7789-20-0
:
Formula:D₂O
MDL Number:MFCD00044636
:20 g/mol
Merck Index:12,02984
Boiling Pt:101.3 °C (1013 hPa)
Melting Pt:3.8 °C
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:232-148-9
Frequently Bought Together