Streptavidin, Xtra™ IF555
Supplier: AAT BIOQUEST
Certificates
About this item
Streptavidin conjugates are widely used in combing with biotin conjugates for detecting a variety of biological targets such as proteins, nucleic acids and other molecules.
- Improved signal
- Improved photostability
- Multiple applications
Caution: For research use only.
Specifications
- Conjugation:Alexa Fluor® 555
- Protein/peptide type:Native
- Species:Human
- Protein synonyms:Avidin
- Protein/peptide name:Streptavidin
Specifications
Frequently Bought Together