Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Dibromobimane [bBBr] ≥90%

Plastic bottle, 5 mg
Catalog # AATB634
Irritant
Supplier:   AAT Bioquest
Dibromobimane [bBBr] ≥90%
Catalog # AATB634
Supplier:  AAT Bioquest
CAS Number:  68654-25-1
Dibromobimane [bBBr] ≥90%
Dibromobimane [bBBr] ≥90%

Plastic bottle, 5 mg

CAS Number:  68654-25-1
Price
£108.00
Log in for availability
1 * 5 mg
Product Details & Documents

dBBr is one of the most popular thiol-reactive fluorescent tags, and is widely used to detect various thiol-containing biomolecules such as glutathione in cells.

  • CAS Number:68654-25-1
  • Formula:C₁₀H₁₀Br₂N₂O₂
  • Molecular Weight:350.01 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg

Specifications